Assay Details
Target Gene Details
Entrez Gene ID: | 475 |
Gene Name: | antioxidant 1 copper chaperone |
Gene Aliases: |
ATX1, HAH1 |
Location: |
Chr.5:151742822-151758649 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATOX1 | NM_004045.3 | NP_004036.1 | ||
| AK128210.1 | 3 | 2367 | ||
| BC112248.1 | AAI12249.1 | |||
| BC112250.1 | AAI12251.1 | |||
| BI599233.1 | ||||
| CN280769.1 |
More Information
Set Membership: |
|

Genomic Map