Assay Details
Target Gene Details
Entrez Gene ID: | 7015 |
Gene Name: | telomerase reverse transcriptase |
Gene Aliases: |
CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TP2, TRT, hEST2, hTRT |
Location: |
Chr.5:1253167-1295626 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TERT | NM_001193376.1 | NP_001180305.1 | ||
| NM_198253.2 | NP_937983.2 | |||
| XM_017009796.1 | XP_016865285.1 | |||
| AB085628.1 | BAC11010.1 | |||
| AB086379.1 | BAC11014.1 | |||
| AB086950.1 | BAC11015.1 | |||
| AF015950.1 | AAC51672.1 | |||
| AF018167.1 | AAC51724.1 | |||
| AX810036.1 | CAE75638.1 | |||
| AX810378.1 | CAE75644.1 | |||
| JF896281.1 | ||||
| JF896283.1 | ||||
| JF896284.1 | ||||
| JF896285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575810 | Chr.5:1055511 - 1282578 on Build GRCh38 | Gain |
|
| nsv950627 | Chr.5:1008886 - 1300685 on Build GRCh38 | Deletion |
|
| nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map