Assay Details
Target Gene Details
Entrez Gene ID: | 4724 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit S4 |
Gene Aliases: |
AQDQ, CI-18, CI-18 kDa, CI-AQDQ |
Location: |
Chr.5:53560610-53683341 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS4 | NM_001318051.1 | NP_001304980.1 | ||
NM_002495.3 | NP_002486.1 | |||
NR_134473.1 | ||||
NR_134474.1 | ||||
NR_134475.1 | ||||
XM_017009491.1 | XP_016864980.1 | |||
AF020351.1 | AAB87865.1 | |||
AK307973.1 | ||||
BC005270.1 | AAH05270.1 | |||
BU145342.1 | ||||
BU954271.1 | ||||
CD692133.1 | ||||
DA609203.1 | ||||
DB489661.1 | ||||
KU178226.1 | ||||
KU178227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5666n100 | Chr.5:53231175 - 54453241 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
