Assay Details
Target Gene Details
Entrez Gene ID: | 4888 |
Gene Name: | neuropeptide Y receptor Y6 (pseudogene) |
Gene Aliases: |
NPY1RL, NPY6RP, PP2, Y2B |
Location: |
Chr.5:137801193-137810751 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY6R | NR_002713.3 | |||
| AK299427.1 | ||||
| DC357138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830500 | Chr.5:137672066 - 137845314 on Build GRCh38 | Loss |
|
| nsv830501 | Chr.5:137788777 - 137977035 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map