Assay Details
Target Gene Details
Entrez Gene ID: | 2676 |
Gene Name: | GDNF family receptor alpha 3 |
Gene Aliases: |
GDNFR3 |
Location: |
Chr.5:138252380-138274564 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFRA3 | NM_001496.3 | NP_001487.2 | ||
AF051767.1 | AAC24355.1 | |||
AK297693.1 | ||||
AK314022.1 | ||||
AY358997.1 | AAQ89356.1 | |||
AY359037.1 | AAQ89396.1 | |||
BC037951.1 | AAH37951.1 | |||
FJ200487.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
