Assay Details
Target Gene Details
| Entrez Gene ID: | 8382 | 
| Gene Name: | NME/NM23 family member 5 | 
| Gene Aliases: | NM23-H5, NM23H5, RSPH23 | 
| Location: | Chr.5:138115172-138139443 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NME5 | NM_003551.2 | NP_003542.1 | ||
| XM_005272099.1 | XP_005272156.1 | |||
| XM_006714716.3 | XP_006714779.1 | |||
| XM_017009945.1 | XP_016865434.1 | |||
| AF067724.1 | AAC64358.1 | |||
| AK312181.1 | ||||
| BC026182.2 | AAH26182.1 | |||
| KU178416.1 | ||||
| KU178417.1 | ||||
| U90450.1 | AAC69440.1 | |||
| Y14992.1 | CAA75226.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  non-DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top