Assay Details
Target Gene Details
Entrez Gene ID: | 51247 |
Gene Name: | poly(A) binding protein interacting protein 2 |
Gene Aliases: |
PAIP-2, PAIP2A |
Location: |
Chr.5:139341832-139369720 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAIP2 | NM_001033112.2 | NP_001028284.1 | ||
| NM_016480.4 | NP_057564.3 | |||
| NR_109830.1 | ||||
| XM_011543428.1 | XP_011541730.1 | |||
| XM_017009522.1 | XP_016865011.1 | |||
| XM_017009523.1 | XP_016865012.1 | |||
| AF151052.1 | AAF36138.1 | |||
| AF317675.1 | AAK11562.1 | |||
| AK024552.1 | ||||
| AK314672.1 | ||||
| AL136640.1 | CAB66575.1 | |||
| BC001716.1 | AAH01716.1 | |||
| BC039337.1 | AAH39337.1 | |||
| BC048106.1 | AAH48106.1 | |||
| BC062718.1 | AAH62718.1 | |||
| BI552800.1 | ||||
| BI560379.1 | ||||
| DB448759.1 |
More Information
Set Membership: |
|

Genomic Map