Assay Details
Target Gene Details
Entrez Gene ID: | 9180 |
Gene Name: | oncostatin M receptor |
Gene Aliases: |
IL-31R-beta, IL-31RB, OSMRB, PLCA1 |
Location: |
Chr.5:38845858-38945579 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSMR | NM_001168355.2 | NP_001161827.1 | ||
| NM_001323504.1 | NP_001310433.1 | |||
| NM_001323505.1 | NP_001310434.1 | |||
| NM_001323506.1 | NP_001310435.1 | |||
| NM_001323507.1 | NP_001310436.1 | |||
| NM_003999.2 | NP_003990.1 | |||
| XM_005248384.1 | XP_005248441.1 | |||
| XM_005248386.2 | XP_005248443.1 | |||
| XM_005248387.2 | XP_005248444.1 | |||
| XM_011514161.2 | XP_011512463.1 | |||
| XM_017010019.1 | XP_016865508.1 | |||
| AI417125.1 | ||||
| AK304016.1 | ||||
| BC010943.1 | AAH10943.1 | |||
| BC063468.1 | AAH63468.1 | |||
| BC125209.1 | ||||
| BC125210.1 | ||||
| DR006275.1 | ||||
| U60805.1 | AAC50946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4801 | Chr.5:38828940 - 38860474 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map