Assay Details
Target Gene Details
Entrez Gene ID: | 10085 |
Gene Name: | EGF like repeats and discoidin domains 3 |
Gene Aliases: |
DEL1 |
Location: |
Chr.5:83940595-84384867 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDIL3 | NM_001278642.1 | NP_001265571.1 | ||
| NM_005711.4 | NP_005702.3 | |||
| AK312858.1 | ||||
| BC030828.1 | AAH30828.1 | |||
| BC053656.1 | AAH53656.1 | |||
| BX648583.1 | ||||
| U70312.1 | AAC02648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830368 | Chr.5:83947456 - 84119801 on Build GRCh38 | Loss |
|
| dgv5709n100 | Chr.5:83955411 - 83987123 on Build GRCh38 | Loss |
|
| nsv1015852 | Chr.5:83404850 - 84499122 on Build GRCh38 | Gain |
|
| nsv525596 | Chr.5:83926497 - 83979269 on Build GRCh38 | Loss |
|
| nsv823108 | Chr.5:83893958 - 84033177 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map