Assay Details
Target Gene Details
Entrez Gene ID: | 105379198 |
Gene Name: | uncharacterized LOC105379198 |
Gene Aliases: |
- |
Location: |
Chr.5:127218484-127229743 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379198 | XM_011543790.2 | XP_011542092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033607 | Chr.5:127049489 - 127968767 on Build GRCh38 | Gain |
|

Genomic Map