Assay Details
Target Gene Details
Entrez Gene ID: | 101929338 |
Gene Name: | uncharacterized LOC101929338 |
Gene Aliases: |
- |
Location: |
Chr.5:9511333-9518086 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTD-2201E9.1 | NR_109935.1 | |||
| AK057486.1 |
Target Gene Details
Entrez Gene ID: | 9037 |
Gene Name: | semaphorin 5A |
Gene Aliases: |
SEMAF, semF |
Location: |
Chr.5:9035026-9546121 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA5A | NM_003966.2 | NP_003957.2 | ||
| XM_006714506.2 | XP_006714569.1 | |||
| XM_011514155.2 | XP_011512457.1 | |||
| XM_011514156.1 | XP_011512458.1 | |||
| XM_011514157.1 | XP_011512459.1 | |||
| XM_011514158.1 | XP_011512460.1 | |||
| XM_017010016.1 | XP_016865505.1 | |||
| AK307587.1 | ||||
| BC115696.1 | AAI15697.1 | |||
| DA336109.1 | ||||
| U52840.1 | AAC09473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575815 | Chr.5:9515067 - 9517330 on Build GRCh38 | Gain |
|
| dgv5565n100 | Chr.5:9218820 - 9591178 on Build GRCh38 | Gain |
|
| nsv1032672 | Chr.5:9182297 - 9843781 on Build GRCh38 | Gain |
|
| nsv597071 | Chr.5:9182264 - 9760487 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map