Assay Details
Target Gene Details
Entrez Gene ID: | 689 |
Gene Name: | basic transcription factor 3 |
Gene Aliases: |
BETA-NAC, BTF3a, BTF3b, NACB |
Location: |
Chr.5:73498425-73505623 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTF3 | NM_001037637.1 | NP_001032726.1 | ||
| NM_001207.4 | NP_001198.2 | |||
| AB062126.1 | BAB93458.1 | |||
| AK225964.1 | ||||
| AK291125.1 | ||||
| BC008062.2 | AAH08062.1 | |||
| BT007120.1 | AAP35784.1 | |||
| BX359147.2 | ||||
| X53280.1 | CAA37375.1 | |||
| X74070.1 | CAA52200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv968944 | Chr.5:73502535 - 73505916 on Build GRCh38 | Duplication |
|
| nsv830346 | Chr.5:73503223 - 73642429 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map