Assay Details
Target Gene Details
Entrez Gene ID: | 64924 |
Gene Name: | solute carrier family 30 member 5 |
Gene Aliases: |
ZNT5, ZNTL1, ZTL1, ZnT-5 |
Location: |
Chr.5:69093949-69131072 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A5 | NM_001251969.1 | NP_001238898.1 | ||
| NM_022902.4 | NP_075053.2 | |||
| NM_024055.4 | NP_076960.1 | |||
| XM_005248569.3 | XP_005248626.1 | |||
| XM_006714672.3 | XP_006714735.1 | |||
| XM_017009749.1 | XP_016865238.1 | |||
| AF461760.1 | AAL96437.1 | |||
| AK292925.1 | ||||
| AY089991.1 | AAM09099.1 | |||
| AY358505.1 | AAQ88869.1 | |||
| BC000808.1 | AAH00808.1 | |||
| BC017441.1 | AAH17441.1 | |||
| BC130452.1 | ||||
| BC130454.1 | ||||
| BC144349.1 | ||||
| BX537394.1 | CAD97636.1 | |||
| DB090660.1 | ||||
| DB259372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map