Assay Details
Target Gene Details
Entrez Gene ID: | 65056 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
GPBP, SSH6, VASCULIN |
Location: |
Chr.5:57173826-57264679 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPBP1 | XM_006714673.3 | XP_006714736.1 | ||
| XM_006714674.3 | XP_006714737.1 | |||
| XM_017009757.1 | XP_016865246.1 | |||
| XM_017009758.1 | XP_016865247.1 | |||
| XM_017009759.1 | XP_016865248.1 |
More Information
Set Membership: |
|

Genomic Map