Assay Details
Target Gene Details
Entrez Gene ID: | 6444 |
Gene Name: | sarcoglycan delta |
Gene Aliases: |
35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD |
Location: |
Chr.5:155728636-156767788 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGCD | XM_017009724.1 | XP_016865213.1 |
More Information
Set Membership: |
|

Genomic Map