Assay Details
Target Gene Details
Entrez Gene ID: | 2107 |
Gene Name: | eukaryotic translation termination factor 1 |
Gene Aliases: |
D5S1995, ERF, ERF1, RF1, SUP45L1, TB3-1 |
Location: |
Chr.5:138506093-138543300 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETF1 | NM_001256302.1 | NP_001243231.1 | ||
| NM_001282185.1 | NP_001269114.1 | |||
| NM_001291974.1 | NP_001278903.1 | |||
| NM_001291975.1 | NP_001278904.1 | |||
| NM_004730.3 | NP_004721.1 | |||
| XM_005271921.1 | XP_005271978.1 | |||
| XM_017009221.1 | XP_016864710.1 | |||
| XM_017009222.1 | XP_016864711.1 | |||
| AK302364.1 | ||||
| AK312510.1 | ||||
| BC014269.2 | AAH14269.1 | |||
| BC088358.1 | AAH88358.1 | |||
| BI460904.1 | ||||
| BM819293.1 | ||||
| DB060174.1 | ||||
| DC297662.1 | ||||
| M75715.1 | AAA36665.1 | |||
| U90176.1 | AAB49726.1 | |||
| X81625.1 | CAA57281.1 |
More Information
Set Membership: |
|

Genomic Map