Assay Details
Target Gene Details
Entrez Gene ID: | 662 |
Gene Name: | BCL2 interacting protein 1 |
Gene Aliases: |
NIP1, SEC20, TRG-8 |
Location: |
Chr.5:173144442-173164387 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BNIP1 | NM_001205.2 | NP_001196.2 | ||
| NM_013978.2 | NP_053581.2 | |||
| NM_013979.2 | NP_053582.2 | |||
| NM_013980.2 | NP_053583.2 | |||
| XM_011534638.1 | XP_011532940.1 | |||
| XM_011534639.1 | XP_011532941.1 | |||
| AF083956.1 | AAC33124.1 | |||
| AF083957.1 | AAC33125.1 | |||
| AF083958.1 | AAC33126.1 | |||
| AU311051.1 | ||||
| AY216799.1 | AAO91805.1 | |||
| BC010959.1 | AAH10959.1 | |||
| U15172.1 | AAC00020.1 |
More Information
Set Membership: |
|

Genomic Map