Assay Details
Target Gene Details
Entrez Gene ID: | 134429 |
Gene Name: | StAR related lipid transfer domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.5:111496033-111512597 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD4 | NM_001308056.1 | NP_001294985.1 | ||
| NM_001308057.1 | NP_001294986.1 | |||
| NM_001308058.1 | NP_001294987.1 | |||
| NM_001308059.1 | NP_001294988.1 | |||
| NM_001308060.1 | NP_001294989.1 | |||
| NM_001308061.1 | NP_001294990.1 | |||
| NM_139164.2 | NP_631903.1 | |||
| NR_131752.1 | ||||
| NR_131753.1 | ||||
| NR_131754.1 | ||||
| XM_017009041.1 | XP_016864530.1 | |||
| XM_017009042.1 | XP_016864531.1 | |||
| XM_017009043.1 | XP_016864532.1 | |||
| XM_017009044.1 | XP_016864533.1 | |||
| AF480299.1 | AAL87129.1 | |||
| AK054566.1 | BAB70759.1 | |||
| AK125317.1 | ||||
| AK290312.1 | ||||
| AK308851.1 | ||||
| AK310848.1 | ||||
| AK315863.1 | ||||
| AK315869.1 | ||||
| AL710253.1 | ||||
| BC042956.1 | AAH42956.1 | |||
| BP234859.1 | ||||
| BP247836.1 | ||||
| DB475661.1 | ||||
| DT220890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950113 | Chr.5:111495303 - 111505002 on Build GRCh38 | Duplication |
|
| nsv830449 | Chr.5:111414293 - 111585511 on Build GRCh38 | Gain+Loss |
|
| nsv518727 | Chr.5:111333002 - 111542854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map