Assay Details
Target Gene Details
Entrez Gene ID: | 1008 |
Gene Name: | cadherin 10 |
Gene Aliases: |
- |
Location: |
Chr.5:24487100-24644978 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH10 | NM_001317222.1 | NP_001304151.1 | ||
| NM_001317224.1 | NP_001304153.1 | |||
| NM_006727.4 | NP_006718.2 | |||
| XM_011513923.2 | XP_011512225.1 | |||
| XM_017008915.1 | XP_016864404.1 | |||
| AB035303.1 | BAA87417.1 | |||
| KJ534806.1 | ||||
| KJ534807.1 | ||||
| KJ534808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830236 | Chr.5:24387686 - 24564620 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map