Assay Details
Target Gene Details
Entrez Gene ID: | 57556 |
Gene Name: | semaphorin 6A |
Gene Aliases: |
HT018, SEMA, SEMA6A1, SEMAQ, VIA |
Location: |
Chr.5:116443555-116574926 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6A | NM_001300780.1 | NP_001287709.1 | ||
| NM_020796.4 | NP_065847.1 | |||
| XM_005272042.3 | XP_005272099.2 | |||
| XM_006714663.1 | XP_006714726.1 | |||
| XM_017009675.1 | XP_016865164.1 | |||
| XM_017009676.1 | XP_016865165.1 | |||
| AB037789.1 | ||||
| AF279656.1 | AAG29378.1 | |||
| AK027439.1 | BAB55111.1 | |||
| AK027471.1 | BAB55136.1 | |||
| AK027501.1 | BAB55158.1 | |||
| AK027654.1 | BAB55269.1 | |||
| AK027867.1 | BAB55418.1 | |||
| AK074975.1 | BAC11326.1 | |||
| AK315143.1 |
Target Gene Details
Entrez Gene ID: | 101927233 |
Gene Name: | SEMA6A antisense RNA 1 |
Gene Aliases: |
CTB-118N6.3 |
Location: |
Chr.5:116447498-116471673 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6A-AS1 | NR_109879.1 | |||
| CD300011.1 |
More Information
Set Membership: |
|

Genomic Map