Assay Details
Target Gene Details
Entrez Gene ID: | 100505678 |
Gene Name: | STARD4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:111512226-111739726 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD4-AS1 | NR_040093.1 | |||
| CR749489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830449 | Chr.5:111414293 - 111585511 on Build GRCh38 | Gain+Loss |
|
| nsv518727 | Chr.5:111333002 - 111542854 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72792094] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map