Assay Details
Target Gene Details
Entrez Gene ID: | 100996485 |
Gene Name: | chromosome 5 open reading frame 66 |
Gene Aliases: |
- |
Location: |
Chr.5:135033280-135344680 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C5orf66 | NM_001277348.1 | NP_001264277.1 | ||
| AK026965.1 | BAB15606.1 |
Target Gene Details
Entrez Gene ID: | 340073 |
Gene Name: | C5orf66 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.5:135236234-135248177 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C5orf66-AS2 | NR_037895.1 | 2 | 99 | |
| AA469978.1 | 1 | 62 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830496 | Chr.5:135205341 - 135340397 on Build GRCh38 | Gain |
|

Genomic Map