Assay Details
Target Gene Details
Entrez Gene ID: | 353376 |
Gene Name: | toll like receptor adaptor molecule 2 |
Gene Aliases: |
MyD88-4, TICAM-2, TIRAP3, TIRP, TRAM |
Location: |
Chr.5:115578496-115602479 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TICAM2 | NM_021649.7 | NP_067681.1 | ||
| AB097022.1 | BAC77375.1 | |||
| AB109098.1 | BAC98399.1 | |||
| BC109265.2 | AAI09266.1 | |||
| BC109266.2 | AAI09267.1 | |||
| BX956265.1 | ||||
| DA992598.1 |
Target Gene Details
Entrez Gene ID: | 100302736 |
Gene Name: | TMED7-TICAM2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.5:115578496-115626179 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED7-TICAM2 | NM_001164468.3 | NP_001157940.1 | ||
| NM_001164469.3 | NP_001157941.1 | |||
| AK075218.1 | BAC11479.1 | |||
| AY275836.1 | AAP81748.1 | |||
| AY304581.1 | AAQ97430.1 | |||
| AY304583.1 | AAQ97432.1 | |||
| AY304584.1 | AAQ97433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830455 | Chr.5:115425845 - 115602587 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map