Assay Details
Target Gene Details
Entrez Gene ID: | 101927908 |
Gene Name: | uncharacterized LOC101927908 |
Gene Aliases: |
- |
Location: |
Chr.5:167164934-167168401 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927908 | NR_134280.1 |
Target Gene Details
Entrez Gene ID: | 57451 |
Gene Name: | teneurin transmembrane protein 2 |
Gene Aliases: |
ODZ2, TEN-M2, TNM2, ten-2 |
Location: |
Chr.5:166979066-168264159 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TENM2 | XM_005265950.2 | XP_005266007.1 | ||
| XM_006714897.2 | XP_006714960.1 | |||
| XM_011534604.2 | XP_011532906.1 | |||
| XM_017009660.1 | XP_016865149.1 | |||
| XM_017009661.1 | XP_016865150.1 | |||
| XM_017009662.1 | XP_016865151.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830545 | Chr.5:167120455 - 167225256 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map