Assay Details
Target Gene Details
Entrez Gene ID: | 3781 |
Gene Name: | potassium calcium-activated channel subfamily N member 2 |
Gene Aliases: |
KCa2.2, SK2, SKCA2, SKCa 2, hSK2 |
Location: |
Chr.5:114056006-114496500 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNN2 | XM_011543387.1 | XP_011541689.1 | ||
| XM_011543389.1 | XP_011541691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471042 | Chr.5:113824793 - 114105195 on Build GRCh38 | Gain |
|
| esv3570362 | Chr.5:114073698 - 114085390 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map