Assay Details
Target Gene Details
Entrez Gene ID: | 105378159 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.6:130288003-130386905 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105378159 | XM_017011599.1 | XP_016867088.1 |
Target Gene Details
Entrez Gene ID: | 154075 |
Gene Name: | sterile alpha motif domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:130133203-130365425 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD3 | NM_001258275.2 | NP_001245204.1 | ||
| AK091351.1 | BAC03644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140417432] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map