Assay Details
Target Gene Details
Entrez Gene ID: | 5467 |
Gene Name: | peroxisome proliferator activated receptor delta |
Gene Aliases: |
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB |
Location: |
Chr.6:35342558-35428191 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830641 | Chr.6:35314757 - 35472826 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map