Assay Details
Target Gene Details
Entrez Gene ID: | 56479 |
Gene Name: | potassium voltage-gated channel subfamily Q member 5 |
Gene Aliases: |
Kv7.5 |
Location: |
Chr.6:72621843-73198851 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ5 | NM_001160130.1 | NP_001153602.1 | ||
| NM_001160132.1 | NP_001153604.1 | |||
| NM_001160133.1 | NP_001153605.1 | |||
| NM_001160134.1 | NP_001153606.1 | |||
| NM_019842.3 | NP_062816.2 | |||
| AF202977.1 | AAF69797.1 | |||
| AF249278.1 | AAF91335.1 | |||
| AF263835.1 | AAF73446.1 | |||
| AK299550.1 | ||||
| AK299741.1 | ||||
| AK316070.1 | ||||
| BC117359.1 | ||||
| BC143554.1 |
Target Gene Details
Entrez Gene ID: | 100873997 |
Gene Name: | KCNQ5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:73134803-73143514 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ5-AS1 | NR_046621.1 | |||
| BM552969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519218 | Chr.6:73135576 - 73153133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map