Assay Details
Target Gene Details
Entrez Gene ID: | 27115 |
Gene Name: | phosphodiesterase 7B |
Gene Aliases: |
bA472E5.1 |
Location: |
Chr.6:135808752-136195571 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE7B | XM_011535755.2 | XP_011534057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830816 | Chr.6:135784196 - 135978659 on Build GRCh38 | Gain |
|
| nsv1023345 | Chr.6:135556122 - 135930860 on Build GRCh38 | Loss |
|
| nsv830814 | Chr.6:135659530 - 135835015 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map