Assay Details
Target Gene Details
Entrez Gene ID: | 57492 |
Gene Name: | AT-rich interaction domain 1B |
Gene Aliases: |
6A3-5, BAF250B, BRIGHT, CSS1, DAN15, ELD/OSA1, MRD12, OSA2, P250R |
Location: |
Chr.6:156777847-157210779 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID1B | NM_017519.2 | NP_059989.2 | ||
| NM_020732.3 | NP_065783.3 | |||
| XM_005267069.4 | XP_005267126.2 | |||
| XM_011535984.2 | XP_011534286.2 | |||
| XM_017011103.1 | XP_016866592.1 | |||
| XM_017011104.1 | XP_016866593.1 | |||
| XM_017011105.1 | XP_016866594.1 | |||
| XM_017011106.1 | XP_016866595.1 | |||
| XM_017011107.1 | XP_016866596.1 | |||
| XM_017011108.1 | XP_016866597.1 | |||
| AF253515.1 | AAN70985.1 | |||
| AF468300.1 | AAL76077.1 | |||
| AF521671.1 | AAN03447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519889 | Chr.6:156751681 - 156788667 on Build GRCh38 | Loss |
|
| nsv823884 | Chr.6:156740673 - 156808901 on Build GRCh38 | Loss |
|
| dgv1071n67 | Chr.6:156774446 - 156781554 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map