Assay Details
Target Gene Details
Entrez Gene ID: | 667 |
Gene Name: | dystonin |
Gene Aliases: |
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2 |
Location: |
Chr.6:56457987-56954671 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Gene Details
Entrez Gene ID: | 101930010 |
Gene Name: | uncharacterized LOC101930010 |
Gene Aliases: |
- |
Location: |
Chr.6:56843986-56864338 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101930010 | NR_125866.1 | |||
| CK819990.1 |
More Information
Set Membership: |
|

Genomic Map