Assay Details
Target Gene Details
Entrez Gene ID: | 85315 |
Gene Name: | progestin and adipoQ receptor family member 8 |
Gene Aliases: |
C6orf33, LMPB1, MPRB |
Location: |
Chr.6:52362128-52407777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAQR8 | NM_133367.4 | NP_588608.1 | ||
AF347029.1 | AAL79777.2 | |||
AK315088.1 | ||||
AY424286.1 | AAR08374.1 | |||
BC030664.2 | AAH30664.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950179 | Chr.6:52385103 - 52400802 on Build GRCh38 | Duplication |
![]() |
esv3608996 | Chr.6:52379078 - 52424700 on Build GRCh38 | Gain |
![]() ![]() |
nsv1025395 | Chr.6:52380188 - 52438563 on Build GRCh38 | Gain |
![]() ![]() |
esv2763551 | Chr.6:52391214 - 52395478 on Build GRCh38 | Loss |
![]() |
nsv1027645 | Chr.6:52376160 - 52417602 on Build GRCh38 | Gain |
![]() |
nsv527443 | Chr.6:52390813 - 52398946 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
