Assay Details
Target Gene Details
Entrez Gene ID: | 285740 |
Gene Name: | PHACTR2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:143554115-143569339 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHACTR2-AS1 | NR_027113.2 | 3 | 1661 | |
| NR_027114.2 | 3 | 2066 | ||
| NR_110148.1 | 5 | 1861 | ||
| NR_110149.1 | 3 | 1403 | ||
| HY183893.1 | 1 | 69 |

Genomic Map