Assay Details
Target Gene Details
Entrez Gene ID: | 101928191 |
Gene Name: | uncharacterized LOC101928191 |
Gene Aliases: |
- |
Location: |
Chr.6:10932317-10954089 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928191 | NR_125851.1 |
Target Gene Details
Entrez Gene ID: | 221711 |
Gene Name: | synaptonemal complex protein 2 like |
Gene Aliases: |
C6orf177, NO145, dJ62D2.1 |
Location: |
Chr.6:10886831-10974309 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYCP2L | NM_001040274.2 | NP_001035364.2 | ||
| AK128130.1 | BAC87288.1 | |||
| AM293674.1 | ||||
| BC012225.1 | AAH12225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519009 | Chr.6:10925570 - 10937871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map