Assay Details
Target Gene Details
Entrez Gene ID: | 285848 |
Gene Name: | patatin like phospholipase domain containing 1 |
Gene Aliases: |
ARCI10, dJ50J22.1 |
Location: |
Chr.6:36242523-36313955 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA1 | NM_001145716.2 | NP_001139188.1 | ||
| NM_173676.2 | NP_775947.2 | |||
| XM_011514520.2 | XP_011512822.1 | |||
| XM_017010778.1 | XP_016866267.1 | |||
| AK096074.1 | BAC04697.1 | |||
| BC103905.1 | AAI03906.1 | |||
| BC103906.1 | AAI03907.1 | |||
| BC103907.1 | AAI03908.1 |
More Information
Set Membership: |
|

Genomic Map