Assay Details
Target Gene Details
Entrez Gene ID: | 5467 |
Gene Name: | peroxisome proliferator activated receptor delta |
Gene Aliases: |
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB |
Location: |
Chr.6:35342558-35428191 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPARD | NM_001171818.1 | NP_001165289.1 | ||
| NM_001171819.1 | NP_001165290.1 | |||
| NM_001171820.1 | NP_001165291.1 | |||
| NM_006238.4 | NP_006229.1 | |||
| NM_177435.2 | NP_803184.1 | |||
| XM_005249193.1 | XP_005249250.1 | |||
| XM_006715120.1 | XP_006715183.1 | |||
| XM_006715123.1 | XP_006715186.1 | |||
| XM_011514707.1 | 3 | 414 | XP_011513009.1 | |
| XM_011514709.1 | 3 | 414 | XP_011513011.1 | |
| XM_011514710.1 | XP_011513012.1 | |||
| XM_017010971.1 | 3 | 414 | XP_016866460.1 | |
| XM_017010972.1 | 3 | 414 | XP_016866461.1 | |
| XM_017010973.1 | XP_016866462.1 | |||
| XM_017010974.1 | XP_016866463.1 | |||
| AB099507.1 | BAC78903.1 | |||
| AK122614.1 | ||||
| AK291661.1 | ||||
| AK296425.1 | ||||
| AK304482.1 | ||||
| AK304878.1 | ||||
| BC002715.2 | AAH02715.1 | |||
| BC007578.1 | AAH07578.1 | |||
| DA192442.1 | ||||
| DA476590.1 | ||||
| L07592.1 | AAA36469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830641 | Chr.6:35314757 - 35472826 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map