Assay Details
Target Gene Details
Entrez Gene ID: | 64094 |
Gene Name: | SPARC related modular calcium binding 2 |
Gene Aliases: |
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1 |
Location: |
Chr.6:168441151-168667994 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMOC2 | NM_001166412.1 | NP_001159884.1 | ||
| NM_022138.2 | NP_071421.1 | |||
| XM_011536065.1 | XP_011534367.1 | |||
| XM_011536066.1 | XP_011534368.1 | |||
| AB014730.1 | BAB20267.1 | |||
| AB014737.1 | BAB20274.1 | |||
| AJ420521.1 | ||||
| AK056700.1 | ||||
| AK314613.1 | ||||
| AL832303.1 | CAI46175.1 | |||
| BC028420.1 | AAH28420.1 | |||
| BC047583.1 | AAH47583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830868 | Chr.6:168333086 - 168526732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map