Assay Details
Target Gene Details
Entrez Gene ID: | 105379393 |
Gene Name: | uncharacterized LOC105379393 |
Gene Aliases: |
- |
Location: |
Chr.8:42051750-42104987 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379393 | NR_136589.1 | |||
| HY030269.1 |

Genomic Map