Assay Details
Target Gene Details
Entrez Gene ID: | 6129 |
Gene Name: | ribosomal protein L7 |
Gene Aliases: |
L7, humL7-1 |
Location: |
Chr.8:73290639-73294494 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL7 | XM_006716463.2 | XP_006716526.1 | ||
| AK291119.1 | ||||
| AK313365.1 | ||||
| X52967.1 | CAA37139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv971665 | Chr.8:73291015 - 73293598 on Build GRCh38 | Duplication |
|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140937357] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map