Assay Details
Target Gene Details
Entrez Gene ID: | 51110 |
Gene Name: | lactamase beta 2 |
Gene Aliases: |
CGI-83 |
Location: |
Chr.8:70637266-70669212 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LACTB2 | NM_016027.2 | NP_057111.1 | ||
| AF151841.1 | AAD34078.1 | |||
| AK222699.1 | BAD96419.1 | |||
| AK290201.1 | ||||
| BC000878.1 | AAH00878.1 |
Target Gene Details
Entrez Gene ID: | 286190 |
Gene Name: | LACTB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:70608577-70663279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LACTB2-AS1 | NR_038881.1 | |||
| AL833348.1 |
More Information
Set Membership: |
|

Genomic Map