Assay Details
Target Gene Details
Entrez Gene ID: | 3084 |
Gene Name: | neuregulin 1 |
Gene Aliases: |
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF |
Location: |
Chr.8:31639222-32771716 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRG1 | NM_001159995.2 | NP_001153467.1 | ||
NM_001159999.2 | NP_001153471.1 | |||
NM_001160001.2 | NP_001153473.1 | |||
NM_001322201.1 | NP_001309130.1 | |||
NM_001322202.1 | NP_001309131.1 | |||
NM_013962.2 | NP_039256.2 | |||
XM_011544512.2 | XP_011542814.1 | |||
XM_017013365.1 | XP_016868854.1 | |||
XM_017013366.1 | XP_016868855.1 | |||
XM_017013367.1 | XP_016868856.1 | |||
XM_017013371.1 | XP_016868860.1 | |||
XM_017013372.1 | XP_016868861.1 | |||
CN603652.1 | ||||
CN603654.1 | ||||
EF372273.1 | ||||
EF372274.1 | ||||
EF372275.1 | ||||
EF372276.1 | ||||
EF372277.1 | ||||
EF517295.1 | ||||
EF517296.1 | ||||
EF517297.1 | ||||
GQ983557.1 | ||||
GQ983558.1 | ||||
L12260.1 | AAB59622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3616802 | Chr.8:31672956 - 31686929 on Build GRCh38 | Gain |
![]() |
esv3891380 | Chr.8:31278415 - 31706814 on Build GRCh38 | Gain |
![]() |
nsv831280 | Chr.8:31675415 - 31853981 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
