Assay Details
Target Gene Details
Entrez Gene ID: | 286190 |
Gene Name: | LACTB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:70608577-70663279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LACTB2-AS1 | NR_038881.1 | |||
| AL833348.1 |

Genomic Map