Assay Details
Target Gene Details
Entrez Gene ID: | 169026 |
Gene Name: | solute carrier family 30 member 8 |
Gene Aliases: |
ZNT8, ZnT-8 |
Location: |
Chr.8:116950273-117176714 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC30A8 | NM_001172811.1 | NP_001166282.1 | ||
NM_001172813.1 | NP_001166284.1 | |||
NM_001172815.2 | NP_001166286.1 | |||
AK298294.1 | ||||
AL713790.1 | CAD28545.1 | |||
EF560713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831434 | Chr.8:116772228 - 116965586 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
