Assay Details
Target Gene Details
Entrez Gene ID: | 51435 |
Gene Name: | scavenger receptor class A member 3 |
Gene Aliases: |
APC7, CSR, CSR1, MSLR1, MSRL1 |
Location: |
Chr.8:27633665-27734027 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARA3 | NM_016240.2 | NP_057324.2 | ||
| NM_182826.1 | NP_878185.1 | |||
| XM_017013535.1 | XP_016869024.1 | |||
| XM_017013536.1 | XP_016869025.1 | |||
| XM_017013537.1 | XP_016869026.1 | |||
| AB007829.1 | BAA86201.1 | |||
| AB007830.1 | BAA86202.1 | |||
| BC060811.1 | AAH60811.1 | |||
| BC075067.3 | AAH75067.1 | |||
| BC075068.2 | AAH75068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610867 | Chr.8:27607002 - 27757379 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map