Assay Details
Target Gene Details
Entrez Gene ID: | 195814 |
Gene Name: | short chain dehydrogenase/reductase family 16C, member 5 |
Gene Aliases: |
EPHD-2, RDH#2, RDH-E2, RDHE2, retSDR2 |
Location: |
Chr.8:56299648-56320776 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SDR16C5 | NM_001318049.1 | NP_001304978.1 | ||
NM_001318050.1 | NP_001304979.1 | |||
NM_138969.3 | NP_620419.2 | |||
XM_011517479.1 | XP_011515781.1 | |||
AB083038.1 | BAB91014.1 | |||
AK057667.1 | BAB71545.1 | |||
AK095159.1 | ||||
AK294634.1 | ||||
AY444559.1 | AAS68535.1 | |||
BC037219.1 | AAH37219.2 | |||
BC064525.1 | AAH64525.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
