Assay Details
Target Gene Details
Entrez Gene ID: | 10656 |
Gene Name: | KH RNA binding domain containing, signal transduction associated 3 |
Gene Aliases: |
Etle, SALP, SLM-2, SLM2, T-STAR, TSTAR, etoile |
Location: |
Chr.8:135457461-135656516 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHDRBS3 | NM_006558.2 | NP_006549.1 | ||
| XM_005250757.2 | XP_005250814.1 | |||
| XM_005250758.3 | XP_005250815.1 | |||
| XM_011516797.2 | XP_011515099.1 | |||
| XM_011516798.1 | XP_011515100.1 | |||
| XM_011516799.1 | XP_011515101.1 | |||
| AF051321.1 | AAC99294.1 | |||
| AF051322.1 | AAC99295.1 | |||
| AF069681.1 | AAC24857.1 | |||
| BC032606.2 | AAH32606.1 | |||
| BC068536.1 | AAH68536.1 |
More Information
Set Membership: |
|

Genomic Map