Assay Details
Target Gene Details
Entrez Gene ID: | 6770 |
Gene Name: | steroidogenic acute regulatory protein |
Gene Aliases: |
STARD1 |
Location: |
Chr.8:38141747-38151265 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAR | NM_000349.2 | NP_000340.2 | ||
| XM_006716392.2 | XP_006716455.1 | |||
| AF035277.1 | AAB88174.1 | |||
| AK090513.1 | ||||
| AK315122.1 | ||||
| AL526126.3 | ||||
| BC010550.1 | AAH10550.1 | |||
| BX647744.1 | ||||
| CR456803.1 | CAG33084.1 | |||
| CR541850.1 | CAG46648.1 | |||
| KU178337.1 | ||||
| KU178338.1 | ||||
| S79669.1 | AAB35726.1 | |||
| U17280.1 | AAC50141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616917 | Chr.8:38078635 - 38160673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map