Assay Details
Target Gene Details
Entrez Gene ID: | 9791 |
Gene Name: | phosphatidylserine synthase 1 |
Gene Aliases: |
LMHD, PSS1, PSSA |
Location: |
Chr.8:96261886-96334552 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTDSS1 | NM_001290225.1 | NP_001277154.1 | ||
| NM_014754.2 | NP_055569.1 | |||
| AK293036.1 | ||||
| BC002376.2 | AAH02376.2 | |||
| BC004192.2 | AAH04192.1 | |||
| BC004390.1 | AAH04390.1 | |||
| D14694.1 | BAA03520.1 | |||
| HY087183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020494 | Chr.8:96188447 - 96286981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map