Assay Details
Target Gene Details
| Entrez Gene ID: | 6788 | 
| Gene Name: | serine/threonine kinase 3 | 
| Gene Aliases: | KRS1, MST2 | 
| Location: | Chr.8:98429677-98944329 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 24 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| STK3 | XM_011517251.1 | XP_011515553.1 | ||
| XM_011517253.2 | XP_011515555.1 | |||
| XM_011517255.2 | XP_011515557.1 | |||
| XM_011517258.1 | XP_011515560.1 | |||
| XM_017013756.1 | XP_016869245.1 | |||
| XM_017013759.1 | XP_016869248.1 | |||
| XM_017013760.1 | XP_016869249.1 | |||
| XM_017013763.1 | XP_016869252.1 | |||
| XM_017013764.1 | XP_016869253.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1034492 | Chr.8:98424113 - 98444288 on Build GRCh38 | Gain |  KCNS2  STK3 | 
| nsv831406 | Chr.8:98278517 - 98469848 on Build GRCh38 | Loss |  NIPAL2  KCNS2  STK3 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top