Assay Details
Target Gene Details
Entrez Gene ID: | 157567 |
Gene Name: | ankyrin repeat domain 46 |
Gene Aliases: |
ANK-S, GENX-115279 |
Location: |
Chr.8:100509752-100559786 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD46 | NM_001270379.1 | NP_001257308.1 | ||
| BC050396.1 | AAH50396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891450 | Chr.8:100436188 - 100519676 on Build GRCh38 | Loss |
|

Genomic Map